Skip to Content
Merck
All Photos(1)

Key Documents

SMB01017

Sigma-Aldrich

Neriifolin

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H46O8
CAS Number:
Molecular Weight:
534.68
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

534.68

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1CC(=CC1=O)[C@@H]2[C@@]3([C@@]([C@H]4[C@@H]([C@@]5([C@@H](C[C@H](CC5)O[C@@H]6O[C@H]([C@@H]([C@H]([C@@H]6O)OC)O)C)CC4)C)CC3)(CC2)O)C

InChI

1S/C30H46O8/c1-16-24(32)26(35-4)25(33)27(37-16)38-19-7-10-28(2)18(14-19)5-6-22-21(28)8-11-29(3)20(9-12-30(22,29)34)17-13-23(31)36-15-17/h13,16,18-22,24-27,32-34H,5-12,14-15H2,1-4H3/t16-,18+,19-,20+,21-,22+,24-,25-,26+,27-,28-,29+,30-/m0/s1

InChI key

VPUNMTHWNSJUOG-BAOINKAISA-N

General description

Neriifolin, a steroid cardiac glycoside, is a bioactive natural compound commonly derived from plants such as Cerbera odollam, Cerbera manghas, Thevetia ahouai and Thevetia peruviana and it is functionally related to a digitoxigenin. Current research suggests that this metabolite acts as an inhibitor and may exhibit diverse biological activities, including anticancer, apoptosis inducing, and anti-inflammatory properties.
Natural product derived from plant source.

Application

Neriifolin is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and drug discovery research.

Biochem/physiol Actions

Neriifolin has a role as a cardiotonic drug, a toxin and a neuroprotective agent.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service