CLS401435
Corning® PTFE-coated magnetic stir bar
L × O.D. 1 in. × 3/8 in.
Synonym(s):
Corning stirrer accessory
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
PTFE-coated
octagonal (with pivot ring)
manufacturer/tradename
Corning 401435
L × O.D.
1 in. × 3/8 in.
Related Categories
General description
These 2 x 3/8 inch (5.08 x 0.95 cm) Corning magnetic stir bars are Polytetrafluorethylene (PTFE)-coated for increased chemical resistance.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)