Skip to Content
Merck
All Photos(1)

Documents

AV46061

Sigma-Aldrich

Anti-BLK antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-B lymphoid tyrosine kinase, Anti-MGC10442

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58 kDa

species reactivity

mouse, human, rat, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BLK(640)

Related Categories

Immunogen

Synthetic peptide directed towards the middle region of human BLK

Application

Anti-BLK antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

BLK, also known as B lymphoid kinase, is a 55kDa tyrosine kinase with SH3, SH2 and catalytic domains that contain consensus sequences of the Src protein tyrosine kinase family. BLK is expressed specifically in the B cell lineage and plays a role in signal transduction pathway that is restricted to B lymphoid cells. Stimulation of resting B-lymphocytes with antibodies to surface immunoglobulin (sIgD or sIgM) induces activation of BLK.

Sequence

Synthetic peptide located within the following region: AVVTKEPIYIVTEYMARGCLLDFLKTDEGSRLSLPRLIDMSAQIAEGMAY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Burkhardt et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(16), 7410-7414 (1991-08-15)
Stimulation of resting B lymphocytes with antibodies to surface immunoglobulin (sIgD or sIgM) induces protein tyrosine phosphorylation, implicating one or more B-cell protein-tyrosine kinases (PTKs) in sIg signal transduction. We have evaluated whether members of the src family of PTKs
S M Dymecki et al.
Science (New York, N.Y.), 247(4940), 332-336 (1990-01-19)
Several pathways of transmembrane signaling in lymphocytes involve protein-tyrosine phosphorylation. With the exception of p56lck, a tyrosine kinase specific to T lymphoid cells that associates with the T cell transmembrane proteins CD4 and CD8, the kinases that function in these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service