Skip to Content
Merck
All Photos(1)

Documents

A5226

Sigma-Aldrich

Angiopoietin-1 human

≥90% (SDS-PAGE), recombinant, expressed in NSO cells

Synonym(s):

Ang-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in NSO cells

Quality Level

Assay

≥90% (SDS-PAGE)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ANGPT1(284)

Biochem/physiol Actions

Ang-1 is a ligand with affinity to the Tie-2 receptor tyrosine kinase, which is expressed in early hematopoietic and endothelial cells. Both Ang-1 and Tie-2 play critical roles in embryonic angiogenesis as well as adult vascular integrity.

Physical form

Lyophilized from a 0.2 μm filtered solution in 50 mM Tris and 0.5 M sodium chloride, pH 6.5, containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured by its ability to inhibit serum deprivation induced apoptosis in HUVE cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service