Skip to Content
Merck
All Photos(2)

Documents

W800002

Sigma-Aldrich

Hexadecanolide

≥98%, stabilized

Synonym(s):

Juniper lactone, Oxacycloheptadecan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O2
CAS Number:
Molecular Weight:
254.41
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
10.047
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥98%

contains

BHT as stabilizer

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

fragrance allergen

no known allergens

Organoleptic

balsamic; sweet; animalic

SMILES string

O=C1OCCCCCCCCCCCCCCC1

InChI

1S/C16H30O2/c17-16-14-12-10-8-6-4-2-1-3-5-7-9-11-13-15-18-16/h1-15H2

InChI key

LOKPJYNMYCVCRM-UHFFFAOYSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service