VT001793
Escherichia coli WDCM 00179 Vitroids™
80-130 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Certified reference material, Microbiological standards, E. coli WDCM 00179 Vitroids™, E. coli WDCM 00179 Vitroids™
About This Item
Recommended Products
grade
certified reference material
Quality Level
Agency
ISO 17034
ISO/IEC 17025
form
solid
shelf life
limited shelf life, expiry date on the label
packaging
package of 10 discs
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
application(s)
environmental
food and beverages
microbiology
storage temp.
−20°C
suitability
Escherichia coli
Escherichia spp.
coliforms
General description
The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms.
These microorganisms are certified according to ISO/IEC 17025 and produced under reproducible conditions compliant with ISO 17034 using authenticated strains from CECT®. These ready-to-use microbiological controls minimize the need for maintaining control strains in the test laboratory and guarantee the use of an authenticated control culture for every quality control test.
Vitroids™ are derived from a traceable culture obtained freeze-dried from CECT®.
Application
Features and Benefits
- Ready-to-use at convenient CFU ranges
- No dilution required
- Quick rehydration in 10 minutes
- 2-3 years of shelf life in a standard -20 °C freezer
- Complete with a comprehensive certificate of analysis
- No maintenance of stock cultures
- No recovery time or pre-enrichment step is necessary
- Robust, efficient, and reliable
- Easy-to-use
- Cost-effective
Other Notes
Other cell culture collections:
- DSM 19683
- NCTC 13167
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service