PHL83843
trans-Coutaric acid
phyproof® Reference Substance
Synonym(s):
trans-p-Coumaroyltartaric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H12O8
CAS Number:
Molecular Weight:
296.23
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
storage temp.
2-8°C
SMILES string
O([C@H]([C@@H](O)C(=O)O)C(=O)O)C(=O)\C=C\c1ccc(cc1)O
InChI
1S/C13H12O8/c14-8-4-1-7(2-5-8)3-6-9(15)21-11(13(19)20)10(16)12(17)18/h1-6,10-11,14,16H,(H,17,18)(H,19,20)/b6-3+/t10-,11-/m1/s1
InChI key
INYJZRKTYXTZHP-NNPIPJJVSA-N
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service