NIST166C
Stainless Steel
NIST® SRM® 166c, low-carbon (AISI 316L)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 100 g
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
A unit of SRM 166c includes a bottle containing ~ 100 g of chips ranging in size from 710 μm (25 mesh) to 75 μm (200 mesh). For more information, please refer to the SDS and COA.
SRM 166C_cert
SRM 166C_SDS
SRM 166C_cert
SRM 166C_SDS
Application
Stainless Steel Standard Reference Material (SRM) is primarily intended for validating chemical and instrumental methods of analysis.
Features and Benefits
- NIST-certified values for carbon are provided, and these values are metrologically traceable to the SI unit of mass fraction, expressed as a percentage (%).
- Expiration details, along with instructions for use, are included on the NIST certificate.
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Carbon (C)
See certificate for values and more details at nist.gov/SRM.
Carbon (C)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service