62634
LST-MUG Broth
suitable for microbiology, NutriSelect® Plus
About This Item
Recommended Products
form
powder
Quality Level
shelf life
limited shelf life, expiry date on the label
composition
dipotassium hydrogen phosphate, 2.75 g/L
lactose, 5 g/L
4-methylumbelliferyl-β-D-glucuronide, 0.1 g/L
potassium dihydrogen phosphate, 2.75 g/L
sodium chloride, 5 g/L
sodium lauryl sulfate, 0.1 g/L
L-tryptophan, 1 g/L
tryptose, 20 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
6.8±0.2 (25 °C)
application(s)
agriculture
environmental
food and beverages
water monitoring
microbiology
suitability
selective and differential for Escherichia coli
selective and differential for coliforms
Application
Preparation Note
Other Notes
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service