Skip to Content
Merck
All Photos(1)

Key Documents

YT310RAHW

Millipore

Millipore Rotary Agitator, 115 V/60 Hz

Synonym(s):

Rotary Agitator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24

AC/DC input

115 V, 60 Hz

Quality Level

shipped in

ambient

General description

If your waste samples contain non-volatile or semi-volatile substances or metals, use this Hazardous Waste Filtration System for saparation, extratction, and filtration. Developed in collaboration with the US Environmental Protection Agency (EPA), the system is designed specifically for the US EPA Extraction Procedure (EP) Toxicity Test and Toxicity Characteristic Leaching Procedure (TCLP). The device separates solid and liquid phases of waste samples for analysis.

Features & Benefits:
•Contains interior holder with PTFE coating to prevent heavy metal contamination
•Allows easy introduction of bulky samples through wide entry port and removable top plate
•Disassembles for easy cleaning

Applications:
US EPA EP Toxicity Test and Toxicity Characteristic Leaching Procedure

YT31042HW Hazardous Waste Filtration System, 142 mm

Replacement Parts:
YY2214257 Hand Knob
YY2004055 1-1/2 in. TC Silicone Gasket
YY2004045 Clamp 1-1/2 in. TC
YY2214265 O-ring (2-251), silicone
YY2214253 O-ring (2-251), PTFE
YT3014257 Underdrain Support, PTFE-coated
YY2214251 Legs with Caps, 3/16 in. wrench
YY2214252 Handwheel Wrench for 142 mm Holder

Accessories:
YY2004057 Gasket, 1-1/2 in., PTFE-coated fluoroelastomer
XX6700035 Tubing for Pressure Use, 1/2 in. I.D.

Membrane Filters:
AP4014250 Glass Fiber filter without Binder, AP40, 142 mm
APFF14250 Glass Fiber Filter without Binder, APFF, 142 mm

YT310RAHW Rotary Agitator

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service