Skip to Content
Merck
All Photos(1)

Key Documents

MDE65027H1

Millipore

Millistak+® Pod Depth Filter

DE65, filtration area 0.027 m², Lab Scale

Synonym(s):

Millistak+ Pod Depth Filter, DE65 media series, 0.027 m2 surface area, 1/4 in. Hose Barb connection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690
NACRES:
NB.24
Pricing and availability is not currently available.

Product Name

Millistak+® Pod Depth Filter, DE65, cellulose, filtration area 0.027 m2, hydrophilic

material

cellulose
filter aid (inorganic)
glass fiber-reinforced polypropylene

Quality Level

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

1 bar max. differential pressure (15 psid) at 80 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)
3.5 bar max. inlet pressure (50 psi) at 25 °C

H

14 cm (5.3 in.)

L

22 cm (8.5 in.)

filtration area

0.027 m2

impurities

<0.25 EU/mL (LAL test, Aqueous extraction)

input

sample type liquid

conductivity

1.52 – 1.94 μS/cm

filter grade

DE65

shipped in

ambient

General description

The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MDE65027H1 comes in a POD format featuring a 14 cm (H) x 22 cm (L) and a filtration area of 0.027 m2.

This DE65 media series delivers optimal performance through a gradient density matri, ideal for primary or secondary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.

The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.

Easy to Use

With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.

Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
product water; post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 5 liters per ft²(50 L/m²) of media surface area

Application

For clarification of serum, plasma, vaccines, cell culture or other fluids.

Features and Benefits

  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Specifications

Filter Grade: DE65

Preparation Note

Sterilization Method
1 autoclave cycle of 60 min @ 123 ºC

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service