Skip to Content
Merck
All Photos(2)

Key Documents

MABE872

Sigma-Aldrich

Anti-Histone H3.3 (H3-3A,H3-3B) Antibody

rat monoclonal, 4H2D7

Synonym(s):

Histone H3.3, H3F3A, H3.3A, H3F3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Histone H3.3 Antibody, clone 4H2D7, clone 4H2D7, 1 mg/mL, from rat

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H2D7, monoclonal

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H3F3B(3021)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

KLH-conjugated linear peptide corresponding to human Histone H3.3.

Application

Immunocytochemistry Analysis: A 1:50 dilution from a represenative lot detected Histone H3.3 in HeLa cells.

Western Blotting Analysis: A representative lot from an independent laboratory detected recombinant Histone H3.3 (Harada, A., et al. (2012). EMBO J. 31(13):2994-3007.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Histone H3.3 in C2C12 cell lysate (Harada, A., et al. (2012). EMBO J. 31(13):2994-3007.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Histone H3.3 in C2C12 cells (Harada, A., et al. (2012). EMBO J. 31(13):2994-3007.).

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Histone H3.3 from C2C12 cell lysate (Harada, A., et al. (2012). EMBO J. 31(13):2994-3007.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-Histone H3.3 Antibody, clone 4H2D7 is validated for use in western blotting, ICC & ChIP for the detection of Histone H3.3.

Quality

Evaluated by Western Blotting in HeLa acid extract.

Western Blotting Analysis: 2 µg/mL of this antibody detected Histone H3.3 in 10 µg of HeLa acid extract.

Target description

~15 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca L Murdaugh et al.
Stem cell reports, 16(8), 2014-2028 (2021-07-10)
Histone variants contribute to the complexity of the chromatin landscape and play an integral role in defining DNA domains and regulating gene expression. The histone H3 variant H3.3 is incorporated into genic elements independent of DNA replication by its chaperone
Guang Yang et al.
Nucleic acids research, 48(6), 3001-3013 (2020-01-23)
Nucleosomal histones are barriers to the DNA repair process particularly at DNA double-strand breaks (DSBs). However, the molecular mechanism by which these histone barriers are removed from the sites of DNA damage remains elusive. Here, we have generated a single
Wenlong Xia et al.
Cell death and differentiation, 24(9), 1548-1563 (2017-05-20)
During the brain development, the process of neural stem cells (NSCs) proliferation and differentiation is precisely regulated. The deficiency in the embryonic brain development will cause serious developmental disorders. Epigenetic modifications play critical roles in controlling proliferation and differentiation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service