MAB8770
Anti-Cytomegalovirus Antibody, pp65, clone 12D10
ascites fluid, clone 12D10, Chemicon®
Synonym(s):
CMV, pp65
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
12D10, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
isotype
IgG1
shipped in
wet ice
Specificity
Reacts with lower matrix protein pp65 of human CMV.
Application
Anti-Cytomegalovirus Antibody, pp65, clone 12D10 is an antibody against Cytomegalovirus for use in IF.
IFA at 1:800 to 1:1600
Optimal working dilutions must be determined by end user.
Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
Ascites fluid.
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeat freeze-thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rachel B Gill et al.
The Journal of general virology, 93(Pt 8), 1743-1755 (2012-05-04)
The UL97 protein kinase is a serine/threonine kinase expressed by human cytomegalovirus (CMV) that phosphorylates ganciclovir. An investigation of the subcellular localization of pUL97 in infected cells indicated that, early in infection, pUL97 localized to focal sites in the nucleus
Role of human cytomegalovirus UL131A in cell type-specific virus entry and release.
Adler, B; Scrivano, L; Ruzcics, Z; Rupp, B; Sinzger, C; Koszinowski, U
The Journal of General Virology null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service