Skip to Content
Merck
All Photos(2)

Key Documents

ABT69

Sigma-Aldrich

Anti-CCDC53 Antibody

from rabbit

Synonym(s):

coiled-coil domain containing 53, Coiled-coil domain-containing protein 53, WASH complex subunit CCDC53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CCDC53(51019)

General description

Coiled-coil domain-containing protein 53 (CCDC53) is one of several components comprising the WASH complex, a complex localized to endosomal surfaces that functions to upregulate actin polymerization through the recruiting and activation of Arp2/3 complex. WASH is a critical component in the fission of tubules (transport intermediates involved in the distribution of endosomes). The WASH complex is composed of CCDC53, F-actin-capping protein subunits alpha and beta, WASH, FAM21, KIAA1033, and KIAA196.

Immunogen

GST-tagged recombinant protein corresponding to human CCDC53.

Application

Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
This Anti-CCDC53 Antibody is validated for use in WB for the detection of CCDC53.
Western Blot Analysis: 2.5 µg/mL from a representative lot detected CCDC53 in 10 µg of mouse brain and PC12 cell lysate.

Quality

Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: 2.5 µg/mL of this antibody detected CCDC53 in 10 µg of HepG2 cell lysate.

Target description

~28 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vojtěch Dostál et al.
Traffic (Copenhagen, Denmark), 24(5), 216-230 (2023-03-31)
The pentameric WASH complex facilitates endosomal protein sorting by activating Arp2/3, which in turn leads to the formation of F-actin patches specifically on the endosomal surface. It is generally accepted that WASH complex attaches to the endosomal membrane via the
Ewan MacDonald et al.
The Journal of cell biology, 217(7), 2549-2564 (2018-06-13)
Transmembrane proteins in the sorting endosome are either recycled to their point of origin or destined for lysosomal degradation. Lysosomal sorting is mediated by interaction of ubiquitylated transmembrane proteins with the endosomal sorting complex required for transport (ESCRT) machinery. In
Benjamin J Tyrrell et al.
Pigment cell & melanoma research, 29(5), 559-571 (2016-07-09)
The five-subunit WASH complex generates actin networks that participate in endocytic trafficking, migration and invasion in various cell types. Loss of one of the two subunits WASH or strumpellin in mice is lethal, but little is known about their role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service