Skip to Content
Merck
All Photos(1)

Key Documents

672315

Sigma-Aldrich

Ursolic Acid

A cell-permeable plant-derived pentacyclic triterpene acid that displays anti-inflammatory, immunomodulatory, and antitumor activities.

Synonym(s):

Ursolic Acid, 3β-Hydroxy-urs-12-en-28-oic acid, CG7, PTP1B Inhibitor VII, PTP Inhibitor XXVII, PTPN2/TCPTP Inhibitor II, SHP2 Inhibitor IV, 3β-Hydroxy-urs-12-en-28-oic acid, CG7, PTPN2/TCPTP Inhibitor II, SHP2 Inhibitor IV, PTP1B Inhibitor VII, PTP Inhibitor XXVII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (GC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMF: 50 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O[C@@H]1C([C@H]2[C@@]([C@@H]3[C@]([C@@]4(CC[C@@]5([C@@H]([C@H]([C@@H](CC5)C)C)C4=CC3)C(=O)O)C)(CC2)C)(CC1)C)(C)C

InChI

1S/C30H48O3/c1-18-10-15-30(25(32)33)17-16-28(6)20(24(30)19(18)2)8-9-22-27(5)13-12-23(31)26(3,4)21(27)11-14-29(22,28)7/h8,18-19,21-24,31H,9-17H2,1-7H3,(H,32,33)/t18-,19+,21+,22-,23+,24+,27+,28-,29-,30+/m1/s1

InChI key

WCGUUGGRBIKTOS-GPOJBZKASA-N

General description

A cell-permeable plant-derived pentacyclic triterpene acid that displays anti-inflammatory, immunomodulatory, and antitumor activities. Shown to inhibit the phosphatase activity of SHP2, PTP1B, and TCPTP (IC50 = 2.73, 3.08 and 3.33 µM, respectively) as well as induce IR autophosphorylation independent of its phosphatase inhibitory activity. Ursolic acid is also reported to inhibit NF-κB and STAT3 pathway activation in cancer cultures, resulting in enhanced sensitivity to chemotherapy agents and apoptotic cell death.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Li, Y., et al. 2010. Int. J. Cancer127, 462.
Pathak, A.K., et al. 2007. Mol. Cancer Res.5, 943.
Jung, S.H., et al. 2007. Biochem. J.403, 243.
Zhang, W., et al. 2006. Biochim. Biophys. Acta1760, 1505.
Shishodia, S., et al. 2003. Cancer Res.63, 4375.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

580.5 °F

Flash Point(C)

304.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service