Skip to Content
Merck
All Photos(2)

Documents

1.46589

Millipore

NaCl Peptone Broth + LT

bottle of, ready-to-use, bottle volume 1000 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type screw cap with septum
ready-to-use

packaging

bottle of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

1000 mL

bottle volume

1000 mL , filling volume

pH

7.0± 1.0

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

components

(Lecithin, Tween)

storage temp.

2-25°C

suitability

nonselective for

General description

Maximum RecoveryDiluent (MRD) with Lecithin and Tween® 80 also known asTryptone Salt Broth is a balanced isotonic strength solution. The combinedprotective effect from peptone(casein) in the diluent with osmotic support ofthe saline (sodium chloride) forms the balanced isotonic strength.
Lecithin and Polysorbate (Tween®) 80, are effective against quaternary ammoniumcompounds, amphoteric surfactants, benzamidines, chlorhexidines and dequadin.Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzylalcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols,phenylethyl alcohols and PHB esters.
The low concentration of peptone prevents microbial multiplication within 1-2hours of diluting the sample.

Application

Maximum Recovery Diluent (MRD) with Lecithin and Tween® 80 is used to recover maximum microbes from test samples. It is used for sample preparation and dilution of stock suspensions of test samples.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They are formulated and tested to meet the pharmacopeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy, and reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service