The particle size, pore size, or surface area has not been determined for this product.
1.12201
Indium(III) oxide
99.5+
Synonym(s):
Indium(III) oxide
About This Item
Recommended Products
Quality Level
form
solid
mp
2000 °C
density
7.18 g/cm3 at 20 °C
storage temp.
no temp limit
SMILES string
[In+3].[In+3].[O-2].[O-2].[O-2]
InChI
1S/2In.3O/q2*+3;3*-2
InChI key
PJXISJQVUVHSOJ-UHFFFAOYSA-N
Related Categories
Analysis Note
Assay (by loss on ignition): ≥ 99.5 %
Fe (Iron): ≤ 0.005 %
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hi, what is the particle size, pore size, and surface area of In2O3 ?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service