Skip to Content
Merck
All Photos(1)

Documents

860469P

Avanti

3-deoxy sphingosine

Avanti Research - A Croda Brand 860469P, powder

Synonym(s):

(R,E)-2-aminooctadec-4-en-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37NO
CAS Number:
Molecular Weight:
283.49
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860469P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860469P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/C[C@@H](N)CO

General description

3-deoxy-sphingosine is a structural analog of the long chain sphingoid base, sphingosine. It has four diastereoisomer, namely (R,E)-,(R,Z)-,(S,E)-,(S,Z)-produced from D- and L-serine.

Application

3-deoxy sphingosine has been used in the synthesis of ceramide analogs.

Packaging

5 mL Amber Glass Screw Cap Vial (860469P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of sphingosine analogues: stereoselective synthesis of 3-deoxysphingosine and cis-isomers.
T Kawate et al.
Chemical & pharmaceutical bulletin, 45(12), 2116-2118 (1998-01-20)
Lukasz Japtok et al.
Allergo journal international, 23(2), 54-59 (2014-01-01)
Sphingolipids are essential molecules of the mammalian epidermis. Keratinocytes generate and secrete huge amounts of ceramide-precursors to the extracellular domain of the stratum corneum, where they are further metabolized to specific ceramide species. The arrangement of ceramides to well-organized lipid
Andrej Kováčik et al.
Scientific reports, 10(1), 3832-3832 (2020-03-04)
Ceramides (Cer) are essential components of the skin permeability barrier. To probe the role of Cer polar head groups involved in the interfacial hydrogen bonding, the N-lignoceroyl sphingosine polar head was modified by removing the hydroxyls in C-1 (1-deoxy-Cer) or
William J Valentine et al.
Methods in molecular biology (Clifton, N.J.), 874, 77-87 (2012-04-25)
Intracellular Ca(2+) mobilization is a useful readout to screen for agonists or antagonists of G-protein -coupled receptors (GPCRs). Here, we describe methods to conduct high-throughput screening of stably or transiently transfected HTC4 cells expressing the individual S1P1-5 receptor subtypes. The
Tania C B Santos et al.
Langmuir : the ACS journal of surfaces and colloids, 36(21), 6007-6016 (2020-05-06)
Compared to the canonical sphingoid backbone of sphingolipids (SLs), atypical long-chain bases (LCBs) lack C1-OH (1-deoxy-LCBs) or C1-CH2OH (1-deoxymethyl-LCBs). In addition, when unsaturated, they present a cis-double bond instead of the canonical  Δ4-5 trans-double bond. These atypical LCBs are directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service