Z545716
Molymod® molecular model set
classroom set, basic organic chemistry
All Photos(2)
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
Recommended Products
feature
no. of atoms 19
no. of bonds 24
packaging
pkg of 6 sets
manufacturer/tradename
Molymod MMP-AT2-6
Related Categories
General description
Our Basic Student Organic Model Set is priced to allow each student to make his or her own models of water, ammonia, methane, carbon dioxide, alcohols, acids, esters, chloro-hydrocarbons, and many other substances. The models are easy to assemble and come complete with clearly illustrated instructions. The individual set is packaged in a plastic storage bag. The package of six sets comes with a plastic storage box and six separate bags.
Contents:
10 White Hydrogen 1-hole
4 Black Carbon 4-hole
2 Red Oxygen 2-hole
3 Green Chlorine 1-hole
1 Blue Nitrogen 4-hole
20 Medium Gray Links
4 Long Flexible Gray Links
Contents:
10 White Hydrogen 1-hole
4 Black Carbon 4-hole
2 Red Oxygen 2-hole
3 Green Chlorine 1-hole
1 Blue Nitrogen 4-hole
20 Medium Gray Links
4 Long Flexible Gray Links
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. May be upgraded or customized with additional model parts, for more info, go to sigma-aldrich.com and enter Molymod in our search engine.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)