Skip to Content
Merck
All Photos(1)

Key Documents

MT51344072

Mettler-Toledo InLab® Science Pro-ISM pH electrode

the electrode for just about any sample

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115604
NACRES:
NB.79

material

glass membrane (A411)
glass shaft

description

MultiPin connector
Reference electrolyte 3 mol/L KCl
Reference system ARGENTHAL, with Ag+ -trap
Temperature probe NTC 30 kOhm
Without cable
built for SevenGo Duo SG23 and SevenGo Duo pro SG68 and SG78

manufacturer/tradename

Mettler-Toledo 51343051

temperature measuring range

0-100 °C

L × diam.

170 mm × 12 mm

pH range

0-12

General description

The Mettler Toledo InLab® Science Pro-ISM pH electrode is breathtaking! With features like built-in temperature probe, bridge electrolyte chamber and movable sleeve junction, this technological masterpiece represents the top of the Mettler Toledo electrode portfolio.

The Mettler Toledo meters have the ability to automatically recognize ISM sensors upon connection, transfer calibration data to and from the sensor etc.

Intelligent sensor management (ISM®)
Integrated NTC 30 kΩ temperature probe
Movable glass sleeve
Double junction reference
Special Mettler Toledo A41 glass membrane
ARGENTHAL and silver ion trap
170 mm shaft length

This electrode can handle the vast majority of all pH applications and offers temperature compensation! Interested in having an electrode that can do everything?
  • Emulsions
  • Dispersions
  • TRIS buffers
  • Oily samples
  • Dairy products
  • Non-aqueous samples
  • Highly viscous samples
  • Samples with low ionic strength
  • Samples with unknown composition

Legal Information

ARGENTHAL is a trademark of Mettler-Toledo, Inc.
ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
MultiPin is a trademark of Mettler-Toledo, Inc.
SevenGo Duo is a trademark of Mettler-Toledo, Inc.
SevenGo Duo pro is a trademark of Mettler-Toledo, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service