Skip to Content
Merck
All Photos(1)

Documents

GF32937059

Stainless Steel - AISI 304 alloy, FeCr18Ni10

mesh, 600 x 600mm, 0.38mm nominal aperture, 0.75mm thickness, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh

Synonym(s):

FE228710

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

description

0.37 (open area)
40 x 40 (wires/inch)

form

(mesh)

manufacturer/tradename

Goodfellow 329-370-59

diam. × thickness

0.25 mm × 0.75 mm , plain weave mesh

size

600 mm × 600 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service