Skip to Content
Merck
All Photos(1)

Key Documents

GF29616419

Havar® - high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn

foil, 50 x 50mm, 0.01mm thickness, as rolled, light tight (lt)

Synonym(s):

CO010230

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

description

light tight (lt)

form

foil (as rolled)

manufacturer/tradename

Goodfellow 296-164-19

size × thickness

50 mm × 50 mm × 0.01 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Havar is a registered trademark of Precision Metals, Inc. AMETEK, or its subsidiaries or affiliates.
Havar is a registered trademark of Precision Metals, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service