933384
Diamond Tip
Retractable Scriber
Synonym(s):
diamond scriber
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Related Categories
General description
EMS techware
Application
Sturdy scribers leave a permanent marking on virtually any surface, including metal, glass, or plastic and widely used throughout the laboratories. This is a high quality chrome pen that features appearance as well as utility. The diamond is mounted in a stainless steel tip and refills are available. The tip is finished at a 60° angle.
To improve your productivity and allow you to view more than one sample at a time
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service