Skip to Content
Merck
All Photos(2)

Key Documents

931225

Sigma-Aldrich

CX73

Umicore

Synonym(s):

Dichlorobis(triphenylphosphine)palladium(II), [Pd(PPh3)2Cl2]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H30Cl2P2Pd
CAS Number:
Molecular Weight:
701.90
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22
form:
powder

Quality Level

form

powder

storage temp.

2-8°C

SMILES string

[Pd]([P](c6ccccc6)(c5ccccc5)c4ccccc4)([P](c3ccccc3)(c2ccccc2)c1ccccc1)(Cl)Cl

InChI

1S/2C18H15P.2ClH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2

Application

CX73 is an organometallic complex. It is an efficient cross-coupling catalyst for C-C coupling reaction, such as Negishi coupling, Suzuki coupling, Sonogashira coupling and Heck coupling reaction. Detection of Bis(triphenylphosphine)palladium(II) dichloride by electrospray ionization quadrupole ion trap mass spectrometry using different imidazolium salts as the charge carrier has been reported. It is employed as catalyst for the Heck reaction medium.

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service