Skip to Content
Merck
All Photos(1)

Key Documents

706337

Sigma-Aldrich

Caffeic acid-13C9

99 atom % 13C, 97% (CP)

Synonym(s):

3,4-Dihydroxycinnamic acid-13C9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C9H8O4
CAS Number:
Molecular Weight:
189.09
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

powder

mol wt

189.00 by atom % calculation

technique(s)

mass spectrometry (MS): suitable

mp

211-213 °C (dec.)

mass shift

M+9

SMILES string

O[13C](=O)\[13CH]=[13CH]\[13c]1[13cH][13cH][13c](O)[13c](O)[13cH]1

InChI

1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2+/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1

InChI key

QAIPRVGONGVQAS-RJKLISAOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service