Skip to Content
Merck
All Photos(2)

Documents

559121

Sigma-Aldrich

Ethyl (2-methoxybenzoyl)acetate

99%

Synonym(s):

3-(2-Methoxyphenyl)-3-oxopropanoic acid ethyl ester, Ethyl (o-methoxybenzoyl)acetate, Ethyl 3-(2-methoxyphenyl)-3-oxopropanoate, Ethyl 3-(2-methoxyphenyl)-3-oxopropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4COCH2CO2CH2H3C
CAS Number:
Molecular Weight:
222.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.5320 (lit.)

bp

259-260 °C (lit.)

density

1.144 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccccc1OC

InChI

1S/C12H14O4/c1-3-16-12(14)8-10(13)9-6-4-5-7-11(9)15-2/h4-7H,3,8H2,1-2H3

InChI key

KROPYAVVJDXRPH-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of N-allyl/propargyl enamine carboxylates
  • Enantioselective ruthenium-catalyzed hydrogenation
  • Hydrosilylation reactions catalyzed by chiral BINAM N-heterocyclic carbene rhodium complex
  • Preparation of potential herbicidal agents

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service