Skip to Content
Merck
All Photos(1)

Documents

47060

Sigma-Aldrich

Fluoride on polymer support

extent of labeling: ~3.0 mmol/g F- loading

Synonym(s):

Amberlite IRA 900 F--form

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: solution phase peptide synthesis

Quality Level

extent of labeling

~3.0 mmol/g F- loading

impurities

≤0.1 mmol/g Cl- (resin)
15-20% water

Related Categories

Other Notes

For a convenient synthesis of alkyl fluorides from alkyl halides or sulfonates ; Heterogenous basic catalyst for C- and O-alkylations, sulfenylations and Michael additions ; deprotection of silyl ethers

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Cardillo et al.
The Journal of Organic Chemistry, 49, 701-701 (1984)
C. Cardillo et al.
Chemistry & Industry, 643-643 (1983)
S. Colonna et al.
Journal of the Chemical Society. Perkin Transactions 1, 2248-2248 (1979)
Elena Chiurchiù et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-19)
Highly functionalized furans are the key scaffolds of many pharmaceuticals and bioactive natural products. Herein, we disclose a new fruitful synthesis of polyfunctionalized furans starting from β-nitroenones and α-functionalized ketones. The protocol involves two steps promoted by solid supported species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service