275816
cis-Bis(benzonitrile)dichloroplatinum(II)
98%
Synonym(s):
cis-Dichlorobis(benzonitrile)platinum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
98%
reaction suitability
core: platinum
reagent type: catalyst
mp
224 °C (dec.) (lit.)
SMILES string
Cl[Pt]Cl.N#Cc1ccccc1.N#Cc2ccccc2
InChI
1S/2C7H5N.2ClH.Pt/c2*8-6-7-4-2-1-3-5-7;;;/h2*1-5H;2*1H;/q;;;;+2/p-2
InChI key
WAJRCRIROYMRKA-UHFFFAOYSA-L
Application
Catalyst for:
- Asymmetric hydroformylation reactions
- Allylation reactions
- Carbene insertion into O-H bonds of alcohols
- Cyclopropanation reactions
- Hydrosilylation reactions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service