Skip to Content
Merck
All Photos(1)

Key Documents

196568

Sigma-Aldrich

2,2-Diphenylethanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CHCH2OH
CAS Number:
Molecular Weight:
198.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

powder

bp

190-192 °C/12 mmHg (lit.)

mp

53-56 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

OCC(c1ccccc1)c2ccccc2

InChI

1S/C14H14O/c15-11-14(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,14-15H,11H2

InChI key

NYLOEXLAXYHOHH-UHFFFAOYSA-N

General description

2,2-Diphenylethanol participates in Meerwein-Ponndorf-Verley type reduction reaction of trans-stilbene oxide with boron triisopropoxide in tetrahydrofuran[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of epoxides with boron triisopropoxide. The Meerwein-Ponndorf-Verley type reduction of boron alkoxides. 2.
Cha JS and Park JH.
Bull. Korean Chem. Soc., 23(10), 1377-1378 (2002)
Xiaoyan Tang et al.
Nature communications, 9(1), 2345-2345 (2018-06-13)
Bacterial poly(3-hydroxybutyrate) (P3HB) is a perfectly isotactic, crystalline material possessing properties suitable for substituting petroleum plastics, but high costs and low volumes of its production are impractical for commodity applications. The chemical synthesis of P3HB via ring-opening polymerization (ROP) of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service