Merck
  • Transesterified sesame (Sesamum indicum L.) seed oil as a biodiesel fuel.
Transesterified sesame (Sesamum indicum L.) seed oil as a biodiesel fuel.
Bioresource technology (2008-01-08)
Abdurrahman Saydut, M Zahir Duz, Canan Kaya, Aylin Beycar Kafadar, Candan Hamamci
ABSTRACT
The sesame (Sesamum indicum L.) oil was extracted from the seeds of the sesame that grows in Diyarbakir, SE Anatolia of Turkey. Sesame seed oil was obtained in 58wt/wt%, by traditional solvent extraction. The methylester of sesame (Sesamum indicum L.) seed oil was prepared by transesterification of the crude oil. Transesterification shows improvement in fuel properties of sesame seed oil. This study supports the production of biodiesel from sesame seed oil as a viable alternative to the diesel fuel.
Sigma-Aldrich
Sesame oil, Antioxidant, delivery vehicle for fat-soluble compounds