Skip to Content
Merck
All Photos(1)

Key Documents

89217

Supelco

Propyl benzoate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CH2CH2CH3
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.501 (lit.)

bp

229.5 °C/766 mmHg (lit.)

density

1.026 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCCOC(=O)c1ccccc1

InChI

1S/C10H12O2/c1-2-8-12-10(11)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

UDEWPOVQBGFNGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service