Skip to Content
Merck
All Photos(1)

Documents

82039-U

Hamilton® PRP-X100 (10 µm) HPLC Columns

L × I.D. 20 mm × 2.1 mm, stainless steel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Hamilton PRP-X100 HPLC Guard Cartridge, 10 μm particle size, L × I.D. 20 mm × 2.1 mm, stainless steel

material

stainless steel

packaging

pack of 5 ea

technique(s)

HPLC: suitable

L × I.D.

20 mm × 2.1 mm

matrix

polymeric particle platform
polymeric

matrix active group

ammonium ion, quaternary phase

particle size

10 μm

pore size

100 Å

operating pH

1-14

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

Related Categories

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service