Skip to Content
Merck
All Photos(1)

Key Documents

502073

Supelco

Quebec Ministry of Environmental Phenol MIx

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    2-Chlorophenol
    3-Chlorophenol
    4-Chlorophenol
    2,4-Dinitrophenol
    2,4-Dimethylphenol
    2,3-Dichlorophenol
    2,4-Dichlorophenol
    2,5-Dichlorophenol
    2,6-Dichlorophenol
    3,4-Dichlorophenol
    3,5-Dichlorophenol
    o-Cresol
    m-Cresol
    p-Cresol
    2-Nitrophenol
    4-Nitrophenol
    2,3,4-Trichlorophenol
    2,3,5-Trichlorophenol
    2,3,6-Trichlorophenol
    2,4,5-Trichlorophenol
    2,4,6-Trichlorophenol
    3,4,5-Trichlorophenol
    2,3,4,5-Tetrachlorophenol
    2,3,4,6-Tetrachlorophenol
    2,3,5,6-Tetrachlorophenol
    2-Methyl-4,6-dinitrophenol
    Pentachlorophenol
    See All (27)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service