Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1349

Sigma-Aldrich

Anti-DIP2B Antibody, clone 2C8, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

DIP2 homolog B, Disco-interacting protein 2 homolog B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C8, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 171.49 kDa
observed mol wt ~172 kDa

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DIP2B(57609)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2C8 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Disco-interacting protein 2 homolog B (DIP2B). It targets an epitope within 17 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal half of human Disco-interacting protein 2 homolog B (DIP2B).

Application

Anti-DIP2B, clone 2C8 ZooMAb, Cat. No. ZRB1349, is a recombinant Rabbit monoclonal antibody that specifically targets Disco-interacting protein 2 homolog B (DIP2B) and has been tested for use in Affinity Binding Assay, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected DIP2B in mouse brain, rat brain, HEK293, and human kidney lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected DIP2B in human kidney tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound DIP2B peptide with a KD of 4.24 x 10-6 an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Disco-interacting protein 2 homolog B (UniProt: Q9P265; also known as DIP2 homolog B, DIP2B) is encoded by the DIP2B (also known as KIAA1463) gene (Gene ID: 57609) in human. DIP2B is a member of the DIP2 family that contains a one highly conserved DNA methyltransferase 1 associated protein 1 (DMAP1)-binding domain (aa 14-31) and two AMP-binding domains (aa 370-822 and 1025-1495). The AMP-binding domains are rich in serine, threonine, and glycine and possess catalytic activity. The DMAP1 has a transcription-repressor activity. DIP2B is moderately expressed in adult brain, placenta, skeletal muscle, heart, kidney, pancreas, lung, spleen and colon. Its expression is reported to be weaker in adult liver, kidney, spleen, and ovary, and in fetal brain and liver. In the brain, its highest expression has been observed in the cerebral cortex and the frontal and parietal lobes. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Winnepenninckx, B., et al. (2007). Am. J. Hum. Gen. 80(2); 221-231).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service