Skip to Content
Merck
All Photos(1)

Key Documents

SML3154

Sigma-Aldrich

Topiroxostat

≥98% (HPLC)

Synonym(s):

4-[3-(4-Pyridinyl)-1H-1,2,4-triazol-5-yl]-2-pyridinecarbonitrile, 4-[5-(pyridin-4-yl)-1H-1,2,4-triazol-3-yl]pyridine-2-carbonitrile, 5-(2-Cyano-4-pyridyl)-3-(4-pyridyl)-1,2,4-triazole, FYX 051, FYX-051

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8N6
CAS Number:
Molecular Weight:
248.24
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

N#CC1=NC=CC(C2=NNC(C3=CC=NC=C3)=N2)=C1

InChI

1S/C13H8N6/c14-8-11-7-10(3-6-16-11)13-17-12(18-19-13)9-1-4-15-5-2-9/h1-7H,(H,17,18,19)

InChI key

UBVZQGOVTLIHLH-UHFFFAOYSA-N

Biochem/physiol Actions

Topiroxostat (FYX-051) is an orally active and potent inhibitor of xanthine oxidoreductase (XOR). It appears that inhibition of XOR by topiroxostat is due to the formation a covalent linkage to molybdenum via oxygen in the hydroxylation reaction intermediate. Also, topiroxostat displayed a potent, non-covalent inhibition of XOR. It is approved for the treatment of gout and hyperuricemia in Japan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service