Skip to Content
Merck
All Photos(1)

Key Documents

SMB01003

Sigma-Aldrich

Cyclopenin

≥90% (LC/MS-ELSD)

Synonym(s):

(-)-Cyclopenin, (-)-Cyclopenine, 4-methyl-3′-phenylspiro[1H-1,4-benzodiazepine-3,2′-oxirane]-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2O3
CAS Number:
Molecular Weight:
294.30
UNSPSC Code:
12352205
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

294.3

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

N1(C3(OC3c4ccccc4)C(=O)Nc2c(cccc2)C1=O)C

InChI

1S/C17H14N2O3/c1-19-15(20)12-9-5-6-10-13(12)18-16(21)17(19)14(22-17)11-7-3-2-4-8-11/h2-10,14H,1H3,(H,18,21)

InChI key

APLKWZASYUZSBL-UHFFFAOYSA-N

General description

Cyclopenin, a benzodiazepine alkaloid, is a natural product commonly available from strains of Penicillum cyclopium, Penicillium aurantiogriseum and Penicillum corymbiferum. Existing research suggests that this Penicillium metabolite exerts various biological activities, including antibacterial, antiviral, antitumor, anti-inflammatory, and neuroprotective properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

According to existing research, Cyclopenin displays selective inhibition against acetylcholinesterase (AChE). Furthermore, it demonstrates anti-inflammatory properties by inhibiting nitric oxide production, NF-κB activation, and the expression of inflammatory factors such as IL-1β, IL-6, and iNOS in brain macrophages. In an Alzheimer′s disease model, Cyclopenin exhibited the ability to rescue learning deficits induced by amyloid-β42 overexpression in flies, indicating its potential as an anti-inflammatory agent for neurodegenerative diseases[1]. Isolated from Penicillium polonicum MCCC3A00951, Cyclopenin displayed potent inhibition of influenza neuraminidase (NA) with an IC50 value of 5.02μM, suggesting its potential as a candidate for anti-influenza NA therapy[2]. Cyclopenin, identified in the fungus Penicillium auratiogriseum Sp-19, demonstrated cytotoxicity against the tsFT210 cell line at a concentration of 3 μg/mL-1, implying its potential as an antitumor agent.(3)Cyclopenin, isolated from Penicillium polonicum MCCC3A00951, exhibited strong inhibition of influenza neuraminidase (NA) with an IC50 value of 5.02μM. This suggests that Cyclopenin has the potential to be developed as an anti-influenza NA candidate.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioactive compounds derived from the marine-derived fungus MCCC3A00951 and their influenza neuraminidase inhibition activity in vitro and in silico
Liu S Z, et al.
Natural product research, 35, 5621-5628 (2021)
Liyan Wang et al.
The Journal of antibiotics, 73(9), 622-629 (2020-03-27)
In the course of screening lipopolysaccharide (LPS)-induced nitric oxide (NO) production inhibitors, two related benzodiazepine derivatives, cyclopenol and cyclopenin, were isolated from the extract of a deep marine-derived fungal strain, Aspergillus sp. SCSIOW2. Cyclopenol and cyclopenin inhibited the LPS-induced formation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service