Skip to Content
Merck
All Photos(2)

Key Documents

SAE0098

Sigma-Aldrich

Lyticase from Arthrobacter luteus

free of DNA contaminants, suitable for Microbiome research, ≥2000 units/mg protein, lyophilized powder

Synonym(s):

(1,3)-β-D-Glucan endohydrolase, 1,3-β-Glucan glucohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

biological source

bacterial (Arthrobacter luteus)

form

lyophilized powder

specific activity

≥2000 units/mg protein

feature

DNA free

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

application(s)

diagnostic assay manufacturing

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Yeast cells are difficult to disrupt because the cell walls may form capsules or resistant spores. DNA can be extracted from yeast by using lysing enzymes such as lyticase, chitinase, zymolase, and gluculase to induce partial spheroplast formation; spheroplasts are subsequently lysed to release DNA. Lyticase is preferred to digest cell walls of yeast and generate spheroplasts from fungi for transformation.

Reported to be useful for lysis of Ashbya, Candida, Debaryomyces, Eremothecium, Endomyces, Hansenula, Hanseniaspora, Kloeckera, Kluyveromyces, Lipomyces, Metschikowia, Pichia, Pullularia, Torulopsis, Saccharomyces, Saccharomycopsis, Saccharomycodes, and Schwanniomyces species.

Purified Lyticase SAE0098 undergoes strict quality control testing to ensure the absence of detectable levels of contaminating DNA using 35 cycles PCR amplification of 16S and 18S rDNA using universal primer sets.

Biochem/physiol Actions

Lyticase hydrolyzes poly-β(1→3)-glucose such as yeast cell wall glucan.

Unit Definition

One unit will produce a ΔA800 of 0.001 per min at pH 7.5 at 25 °C, using a suspension of yeast as substrate in a 3 mL reaction mixture.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Enzymatic cell lysis and protoplast prep processes demystified, revealing complex enzymatic reactions during sample preparation.

Enzymatic cell lysis and protoplast prep processes demystified, revealing complex enzymatic reactions during sample preparation.

Enzymatic cell lysis and protoplast prep processes demystified, revealing complex enzymatic reactions during sample preparation.

Enzymatic cell lysis and protoplast prep processes demystified, revealing complex enzymatic reactions during sample preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service