Skip to Content
Merck
All Photos(2)

Documents

SAB4500412

Sigma-Aldrich

Anti-COX41 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

COX IV-1, COX4, COXIV, cytochrome c oxidase polypeptide IV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 19 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COX4I1(1327)

General description

Anti-COX41 Antibody detects endogenous levels of total COX41 protein.
COX41 (cytochrome c oxidase subunit 4I1) is located on human chromosome 16q24. COX4NB (COX4 neighbour) and COX4I1 (COX4 isoform 1) belongs to the cytochrome C oxidase subunit IV (COX4) family.

Immunogen

The antiserum was produced against synthesized peptide derived from human COX41.

Immunogen Range: 11-60

Biochem/physiol Actions

COX4 isoform 1 (COX4-1) (COX4-1) may play an important role in glioma chemoresistance. It regulates the upregulation of BMI1 (polycomb complex protein BMI1). It is necessary for the multiplication of GBM (glioblastoma multiforme). In cells, expression of COX4-1 is necessary to maintain the stability of mitochondria and ROS (reactive oxygen species) homeostasis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A small de novo 16q24.1 duplication in a woman with severe clinical features
Quemener-Redon S, et al.
European Journal of Medical Genetics, 56(4), 211-215 (2013)
Fan Yang et al.
Journal of cellular biochemistry, 119(2), 2427-2439 (2017-09-16)
Mitochondrial dysfunction has emerged as a critical pathophysiological factor of myocardial ischemia/reperfusion (I/R) injury. A thorough understanding of mitochondrial dysfunction during I/R at the molecular level is urgently needed. One prominent microRNA, miR-410, was previously reported to be dynamically regulated
Claudia R Oliva et al.
Oncotarget, 6(6), 4330-4344 (2015-03-03)
Nuclear-encoded cytochrome c oxidase subunit 4 (COX4) is a key regulatory subunit of mammalian cytochrome c oxidase, and recent studies have demonstrated that COX4 isoform 1 (COX4-1) could have a role in glioma chemoresistance. The Polycomb complex protein BMI1 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service