Key Documents
HS234525C
Parafilm® Sealing Film Dispenser, Blue
green (dispenser)
About This Item
UNSPSC Code:
41123300
NACRES:
NB.51
material
acrylic
green (dispenser)
feature
2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS234525C
L × W × H
120 mm × 156 mm × 171 mm
General description
The ABS dispenser has a unique design for maximum safety, storage, dispensing, and cutting Parafilm, tape, labels, and Tough-Tags.
Features and Benefits
  • ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to fingers.
  • Supplied with two easily exchanged cutting blades
  • Dispenser holds one or two 50 mm (2-inch) rolls, or one 100 mm (4-inch) roll
Legal Information
Parafilm is a registered trademark of Bemis Company, Inc.