Skip to Content
Merck
All Photos(1)

Key Documents

GERPN82D

Hybond® ECL nitrocellulose membranes

diam. 82 mm, disks nitrocellulose, pack of 50 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

material

disks nitrocellulose
nitrocellulose

form

solid

feature

wetted part: no

packaging

pack of 50 ea

manufacturer/tradename

Cytiva RPN82D

diam.

82 mm

color

White

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

General description

For all labeling and detection systems. Expect excellent sensitivity, resolution and low background.

Hybond ECL gives excellent sensitivity, resolution, and low background for all labeling and detection systems (radioactive, nonradioactive, and chromogenic), especially when used with ECL Western blotting system, Amersham ECL Prime, Amersham ECL Select, and ECL Plex Western blotting detection system.

Special packaging features include interleaf on both sides of the membrane and an air-tight resealable aluminum bag with label space for marking the "opened on" date.

Features and Benefits

  • 0.45 µm pore size.Special packaging features enhance membrane performance in terms of handling and consistency of results.Unsupported, 100% pure nitrocellulose membrane validated for use with all ECL Western blotting systems and for all protein blotting applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

ECL is a trademark of Cytiva
Hybond is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service