Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1332-01

Repel-silane ES

Cytiva 17-1332-01, pack of 500 mL

Synonym(s):

Silane Treatment

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.25

form

liquid

feature

wetted part: no

packaging

pack of 500 mL

manufacturer/tradename

Cytiva 17-1332-01

storage condition

dry at room temperature
protect from light

color

Colorless

solubility

cold water: soluble (Soluble in cold water and hot water)

General description

PlusOne Repel-Silane ES. Repel-Silane ES is a 2% solution of dimethyldichlorosilane dissolved in octamethylcyclooctasilane, a low toxicity, environmentally safe solvent.

Application

It is used to inhibit sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F

Flash Point(C)

56 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service