Skip to Content
Merck
All Photos(1)

Key Documents

Detroit 510

90102531, human skin, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human skin
growth mode:
Adherent
karyotype:
2n = 46
morphology:
Fibroblast
products:
Not specified
receptors:
Not specified

Product Name

Detroit 510, 90102531

biological source

human skin

growth mode

Adherent

karyotype

2n = 46

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian skin Galactosemia

Cell Line Description

Derived from skin tissue of a 9 month old Caucasian female with Galactosemia. Galactose-1-phosphate transferase negative.

Application

Virus studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 11,12
D16S539: 11,12
D5S818: 10,12
D7S820: 8,12
THO1: 7,9.3
TPOX: 8
vWA: 15,17

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 0.1% Lactalbumin hydrolysate (LH) + 10%Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Finite lifespan, up to 30 population doublings are expected.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service