TOKUNAGA human cell line
IHW Number: 9304, 85102916
Synonym(s):
TOK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human blood
ethnicity
Not specified
gender
male
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblastoid
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
Looking for similar products? Visit Product Comparison Guide
General description
Homozygous: Yes
Cell Line Description
This cell line is an Epstein-Barr Virus (EBV) transformed lymphoblastoid line and is part of the Human Leukocyte Antigen (HLA) Typed Collection maintained by the European Collection of Cell Cultures (ECACC). HLA-Typing Data is available for this cell line.
DNA Profile
A: 2402
HLA DRB1: 1502
HLA DRB5: 102
HLA DRB6: 2
HLA DQA1: 103
HLA DQB1: 601
HLA DPA1: 2
HLA DPB1: 901
HLA DRB1: 1502
HLA DRB5: 102
HLA DRB6: 2
HLA DQA1: 103
HLA DQB1: 601
HLA DPA1: 2
HLA DPB1: 901
Serological Profile
HLA A: 24
HLA B: 52
Bw4/bw6: 4
HLA Dw: 12
HLA DR: 15
HLA DR51: 51
HLA DQ: 6
HLA B: 52
Bw4/bw6: 4
HLA Dw: 12
HLA DR: 15
HLA DR51: 51
HLA DQ: 6
Culture Medium
Subculture Routine
Maintain cultures between 3x105 - 2x106 cells/ml, 5% CO2, 37 °C. Upon resuscitation remove cryoprotectant - centrifuge at 300 x g for 5 minutes and re-suspend the cell pellet in fresh culture medium.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has restrictions and can only be supplied to academic and not-for-profit organisations. Nucleic acids can be supplied to all researchers including those from commercial organisations.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service