Skip to Content
Merck
All Photos(1)

Key Documents

AXYSCT15MLRS

Centrifuge tube with screw cap, conical bottom, co-polymer

capacity 15 mL, units of 25, sterile

Synonym(s):

15 ml centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.22
Pricing and availability is not currently available.

material

conical bottom tube
polypropylene
screw top cap

sterility

sterile

packaging

units of 25

manufacturer/tradename

Axygen Inc SCT-15ML-R-S
Corning

parameter

12000 × g max. RCF

capacity

15 mL

color

clear

fitting

cap (screw cap)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service