Skip to Content
Merck
All Photos(1)

Key Documents

A2980

Sigma-Aldrich

Anti-AcV5 antibody, Mouse monoclonal

clone ACV5, purified from hybridoma cell culture

Synonym(s):

Anti-Baculovirus envelope gp64 protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ACV5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~58 kDa

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 0.5-1 μg/mL using extracts of Sf9 cells infected with baculovirus

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-AcV5 antibody, Mouse monoclonal, (mouse IgG2b isotype) is derived from the hybridoma ACV5 produced by the fusion of mouse myeloma cells and splenocytes from C57Bl/6 X Balb/c mice immunized with AcNPV extracellular nonoccluded virus (NOV). GP64 (AcV5) is an envelope glycoprotein found in the baculovirus Autographa californica multiple nucleopolyhedrovirus. GP64 gene open reading frame encodes a protein of 509 amino acids with a molecular weight of 58 kDa. The protein contains N-linked glycosylation sites and hydrophobic N- and C-termini, characteristic of signal and membrane anchor motifs found in envelope glycoproteins. The GP64 protein is present early (6 hr) post infection and accumulates in the infected cell moving to the periphery later.
gp64 (AcV5) is an envelope glycoprotein found in the baculovirus Autographa californica multiple nucleopolyhedrovirus. It is involved in involved in viral entry into cells by receptor-mediated endocytosis. Elevated expression of gp64 has been shown to enhance gene transfer in a variety of cell types.

Immunogen

AcNPV extracellular nonocculeded virus (NOV).

Application

Anti-AcV5 antibody, Mouse monoclonal has been used in:
  • immunoblotting
  • immunoprecipitation
  • dot blot
  • immunocytochemistry

Proteins tagged with this AcV5 peptide are functional in both dicot and monocot plants and show no cross reactivity across many plant species. Epitope tagged fusion proteins are widely used in protein analysis of biological samples (e.g. plant cell extracts). Antibodies specific for the tag are a valuable means for the detection of the protein. Anti-AcV5 antibody, Mouse monoclonal, clone ACV5 may be used in immunoblotting, immunoprecipitation, dot blot, and immunocytochemistry.

Biochem/physiol Actions

GP64 (AcV5) is involved in viral entry into cells by receptor-mediated endocytosis. Elevated expression of gp64 has been shown to enhance gene transfer in a variety of cell types. AcV5 is a monoclonal antibody that recognizes a nine amino acid residue tag (SWKDASGWS) of baculovirus Autographa californica GP64 envelope fusion protein (efp).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shudi Zhao et al.
Frontiers in immunology, 13, 845268-845268 (2022-03-08)
Antiviral immunity involves various mechanisms and responses, including the RNA interference (RNAi) pathway. During long-term coevolution, viruses have gained the ability to evade this defense by encoding viral suppressors of RNAi (VSRs). It was reported that p35 of baculovirus can
Boris F Mayer et al.
Plant physiology, 182(2), 1022-1038 (2019-12-18)
Anthropogenic climate change precipitates the need to understand plant adaptation. Crucial in temperate climates, adaptation to winter is characterized by cold acclimation and vernalization, which respectively lead to freezing tolerance and flowering competence. However, the progression of these responses during
Autographa californica multiple nucleopolyhedrovirus GP64 protein: roles of histidine residues in triggering membrane fusion and fusion pore expansion
Li Z and Blissard GW
Journal of virology, 85(23), 12492-12504 (2011)
Location, sequence, transcriptional mapping, and temporal expression of the gp64 envelope glycoprotein gene of the Orgyia pseudotsugata multicapsid nuclear polyhedrosis virus
Blissard GW and Rohrmann GF
Virology, 170(2), 537-555 (1989)
Baculovirus GP64-mediated entry into mammalian cells
Kataoka C, et al.
Journal of virology, 86(5), 2610-2620 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service