Skip to Content
Merck
All Photos(1)

Key Documents

16646

Sigma-Aldrich

1,4-Bis[3-(2-pyridyldithio)propionamido]butane

≥85% (HPLC)

Synonym(s):

1,16-Di(2-pyridyl)-1,2,15,16-tetrathia-6,11-diazahexadecane-5,12-dione, DPDPB, N, N′-Tetramethylenebis[3-(2-pyridyldithio)propionamide]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N4O2S4
CAS Number:
Molecular Weight:
482.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥85% (HPLC)

reaction suitability

reagent type: cross-linking reagent

storage temp.

2-8°C

SMILES string

O=C(CCSSc1ccccn1)NCCCCNC(=O)CCSSc2ccccn2

InChI

1S/C20H26N4O2S4/c25-17(9-15-27-29-19-7-1-3-13-23-19)21-11-5-6-12-22-18(26)10-16-28-30-20-8-2-4-14-24-20/h1-4,7-8,13-14H,5-6,9-12,15-16H2,(H,21,25)(H,22,26)

InChI key

JMUAKWNHKQBPGJ-UHFFFAOYSA-N

Application

1,4-Bis[3-(2-pyridyldithio)propionamido]butane can be used to study protein structural analysis, protein modification, proteomics and homobifunctional cross-linking reagents.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Reagent for protein-protein cross-linking via cysteine crosslinking

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christoph Schell et al.
Developmental cell, 47(6), 741-757 (2018-12-07)
Podocytes, highly specialized epithelial cells, build the outer part of the kidney filtration barrier and withstand high mechanical forces through a complex network of cellular protrusions. Here, we show that Arp2/3-dependent actin polymerization controls actomyosin contractility and focal adhesion maturation
G N Zecherle et al.
The Journal of biological chemistry, 267(9), 5889-5896 (1992-03-25)
Oligonucleotide-directed mutagenesis was used to produce a serine 89 to cysteine 89 substitution in the C-terminal globular domain of Escherichia coli ribosomal protein L7/L12. Cys-89 represented the only cysteine residue in the protein. L7/L12Cys89 was overproduced in E. coli and
Sanja Kovačević et al.
Neuroendocrinology, 108(4), 278-290 (2018-12-21)
Increased fructose consumption and chronic exposure to stress have been associated with the development of obesity and insulin resistance. In the hypothalamus, a crossroad of stress responses and energy balance, insulin and glucocorticoids regulate the expression of orexigenic neuropeptides, neuropeptide
Hua Su et al.
Molecular cell, 67(6), 907-921 (2017-08-29)
The class III phosphoinositide 3-kinase VPS34 plays a key role in the regulation of vesicular trafficking and macroautophagy. So far, we know little about the molecular mechanism of VPS34 activation besides its interaction with regulatory proteins to form complexes. Here
Zhenhuan Guo et al.
Science signaling, 7(354), rs7-rs7 (2014-12-04)
E-cadherin-mediated cell-cell adhesion and signaling plays an essential role in development and maintenance of healthy epithelial tissues. Adhesiveness mediated by E-cadherin is conferred by its extracellular cadherin domains and is regulated by an assembly of intracellular adaptors and enzymes associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service