Skip to Content
Merck
All Photos(1)

Key Documents

PHR2725

Supelco

Tilmicosin

certified reference material, pharmaceutical secondary standard

Synonym(s):

20-Deoxo-20-(3,5-dimethyl-1-piperidinyl)desmycosin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H80N2O13
Molecular Weight:
869.13
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1667370

description

Pharmaceutical Secondary Standard; Certified Reference Material

API family

tilmicosin

packaging

pkg of 500 mg

SMILES string

CC1OC(OCC(C(CC)O2)/C=C(/C=C/C(C(C)CC(CCN3CC(C)CC(C)C3)C(OC4OC(C)C(O)C(N(C)C)C4O)C(C)C(O)CC2=O)=O)C)C(OC)C(OC)C1O

InChI

1S/C46H80N2O13/c1-13-36-33(24-57-46-44(56-12)43(55-11)40(53)31(8)59-46)19-25(2)14-15-34(49)28(5)20-32(16-17-48-22-26(3)18-27(4)23-48)42(29(6)35(50)21-37(51)60-36)61-45-41(54)38(47(9)10)39(52)30(7)58-45/h14-15,19,26-33,35-36,38-46,50,52-54H,13,16-18,20-24H2,1-12H3/b15-14+,25-19+/t26-,27+,28-,29+,30-,31-,32+,33-,35-,36-,38+,39-,40-,41-,42-,43-,44-,45+,46-/m1/s1

InChI key

JTSDBFGMPLKDCD-XVFHVFLVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Cardio-vascular system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service