Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8447

Perfluorinated sulfonic acids in methanol

NIST® RM 8447

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.24

grade

certified reference material

Quality Level

manufacturer/tradename

NIST®

format

multi-component solution

Other Notes

Reference/Informational value is provided for the following analytes.
Perfluorobutane sulfonate (C4HF9O3S), Perfluorohexane sulfonate (C6HF13O3S), Perfluorooctane sulfonate (C8HF17O3S)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service