NISTRM8182
Fatty acid Methyl Esters in 2,2,4-Trimethylpentane
NIST®SRM®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
packaging
pkg of 6 mg
manufacturer/tradename
NIST®
technique(s)
gas chromatography (GC): suitable
format
multi-component solution
storage temp.
2-8°C
General description
Other Notes
Reference/Informational value is provided for the following analytes.
Octanoic Acid Methyl Ester, Decanoic Acid Methyl Ester, Dodecanoic Acid Methyl Ester, Tetradecanoic Acid Methyl Ester, Hexadecanoic Acid Methyl Ester, Octadecanoic Acid Methyl Ester, Eicosanoic Acid Methyl Ester, Docosanoic Acid Methyl Ester, Tetracosanoic Acid Methyl Ester, 9-Tetradecenoic Acid Methyl Ester, 9-Hexadecenoic Acid Methyl Ester, 9-Octadecenoic Acid Methyl Ester, 9-trans-Octadecenoic Acid Methyl Ester, 11-Octadecenoic Acid Methyl Ester, 11-trans-Octadecenoic Acid Methyl Ester, 9,12-Octadecadienoic Acid Methyl Ester, 9-trans,12-trans-Octadecadienoic Acid Methyl Ester, 9,12,15-Octadecatrienoic Acid Methyl Ester, 6,9,12-Octadecatrienoic Acid Methyl Ester, 11-Eicosenoic Acid Methyl Ester, 5,8,11,14-Eicosatetraenoic Acid Methyl Ester, 5,8,11,14,17-Eicosapentaenoic Acid Methyl Ester,
7,10,13,16,19-Docosapentaenoic Acid Methyl Ester, 4,7,10,13,16,19-Docosahexaenoic Acid Methyl Ester, 13-Docosenoic Acid Methyl Ester, 15-Tetracosenoic Acid Methyl Ester
See certificate for values and more details at nist.gov/SRM.
Octanoic Acid Methyl Ester, Decanoic Acid Methyl Ester, Dodecanoic Acid Methyl Ester, Tetradecanoic Acid Methyl Ester, Hexadecanoic Acid Methyl Ester, Octadecanoic Acid Methyl Ester, Eicosanoic Acid Methyl Ester, Docosanoic Acid Methyl Ester, Tetracosanoic Acid Methyl Ester, 9-Tetradecenoic Acid Methyl Ester, 9-Hexadecenoic Acid Methyl Ester, 9-Octadecenoic Acid Methyl Ester, 9-trans-Octadecenoic Acid Methyl Ester, 11-Octadecenoic Acid Methyl Ester, 11-trans-Octadecenoic Acid Methyl Ester, 9,12-Octadecadienoic Acid Methyl Ester, 9-trans,12-trans-Octadecadienoic Acid Methyl Ester, 9,12,15-Octadecatrienoic Acid Methyl Ester, 6,9,12-Octadecatrienoic Acid Methyl Ester, 11-Eicosenoic Acid Methyl Ester, 5,8,11,14-Eicosatetraenoic Acid Methyl Ester, 5,8,11,14,17-Eicosapentaenoic Acid Methyl Ester,
7,10,13,16,19-Docosapentaenoic Acid Methyl Ester, 4,7,10,13,16,19-Docosahexaenoic Acid Methyl Ester, 13-Docosenoic Acid Methyl Ester, 15-Tetracosenoic Acid Methyl Ester
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
10.4 °F - closed cup
Flash Point(C)
-12 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service