Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Silicon (Si), Iron (Fe), Nickel (Ni), Magnesium (Mg), Copper (Cu), Manganese (Mn), Titanium (Ti), Zinc (Zn), Chromium (Cr), Lead (Pb), Tin (Sn), Gallium (Ga), Vanadium (V)
See certificate for values and more details at nist.gov/SRM.
Silicon (Si), Iron (Fe), Nickel (Ni), Magnesium (Mg), Copper (Cu), Manganese (Mn), Titanium (Ti), Zinc (Zn), Chromium (Cr), Lead (Pb), Tin (Sn), Gallium (Ga), Vanadium (V)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service